Learn More
Selleck Chemical LLC ONO-AE3-208 E2986-5MG
SDP
Supplier:  Selleck Chemical LLC E29865MG
Encompass_Preferred
ONO-AE3-208(AE 3-208) is a selective and orally active EP4 receptor antagonist with a Ki of 1 3 nM less potent to EP3 FP and TP receptors with Ki of 30 nM 790 nM and 2400 nM respectively
Catalog No. 50-000-40732