Learn More
TARGETMOL CHEMICALS INC ONO-RS-082 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2181510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ONO-RS-082 is a Ca2+-independent phospholipase A2 inhibitor. purity: 99%
Catalog No. 50-257-7204