Learn More
Medchemexpress LLC Ononetin | 487-49-0 | MFCD00020119 | 99.5% | 258.27 g·mol⁻1 | C15H14O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1084515MG
Encompass_Preferred
Ononetin is a natural deoxybenzoin compound that potently and selectively blocks the TRPM3 ion channel (IC50 = 0.3 μM). Supplied for research use, it is provided with high reported purity and is available as a small-mass solid or as a 10 mM DMSO solution for laboratory assays and pharmacological studies.
  • Potent and selective TRPM3 channel blocker (IC50 = 0.3 μM).
  • High reported purity (99.54%).
  • Available as solid (5 mg, 10 mg, 25 mg) and 10 mM solution in DMSO.
  • Suitable for ion-channel pharmacology and TRP channel research.
  • Provided with product datasheet and SDS for laboratory safety and handling.
Catalog No. 50-002-63668