Learn More
Apexbio Technology LLC ONX-0914 (PR-957) 960374-59-8 100mg
SDP
Supplier:  Apexbio Technology LLC A4011100
Encompass_Preferred
ONX-0914 (PR-957) is a selective inhibitor targeting the immunoproteasome subunit 5i (LMP7) a proteolytic component involved in antigenic peptide processing for MHC class I presentation By reversibly binding the 5i-specific catalytic pocket ONX-0914 impairs the production of pro-inflammatory cytokines in activated human peripheral blood mononuclear cells (PBMCs) and suppresses IL-17 secretion during Th17 cell differentiation ONX-0914 has been employed experimentally in preclinical models of autoimmune conditions such as arthritis colitis and diabetes Reported IC50 values are approximately 5 15 nM for the 5i subunit demonstrating selective preference over constitutive proteasome subunits
Catalog No. 50-000-64380