Learn More
Apexbio Technology LLC ONX-0914 (PR-957), 5mg Cas No: 960374-59-8 MFCD: MFCD26794217
SDP
Supplier:  Apexbio Technology LLC A40115
Encompass_Preferred
ONX-0914, previously known as PR-975, is a potent inhibitor of immunoproteasome, a form of proteasome generating peptides presented on major histocompatibility complex (MHC) class I molecule to cytotixic T cells. There are other sizes available. Please inqury us for quote.
Catalog No. NC1566878