Learn More
Medchemexpress LLC Onx-0914 (TFA) | 00-00-0 | 98.0% | C33H41F3N4O9 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13207A10MG
Encompass_Preferred
ONX-0914 TFA is a selective, irreversible inhibitor of the immunoproteasome LMP7 subunit (chymotrypsin-like activity), supplied as the trifluoroacetate salt for research use. It blocks cytokine production and is used to study proteasome function and immune-related disease models. Provided as a high-purity small-molecule reagent.
Catalog No. 50-002-61155