Learn More
Phenomenex Inc Phenomenex Onyx Monolithic C18 LC Column, 100 x 4.6 mm
Supplier:  Phenomenex Inc CH07643
Encompass
The Phenomenex Onyx Monolithic C18 LC column uses a monolithic silica rod stationary phase for rapid reversed-phase separations of hydrophobic compounds, and is designed to perform well with dirty or viscous sample matrices.
  • Monolithic C18 stationary phase for reversed-phase separations.
  • Monolithic silica rod support to provide high permeability and low backpressure.
  • 100 x 4.6 mm analytical column format for fast analytical workflows.
  • Optimized for fast separation of hydrophobic compounds in challenging samples.
  • Supplied as a single column (each) for analytical use.
Catalog No. NC3736790