Learn More
Medchemexpress LLC OP-5244 | 2381268-71-7 | 99.1% | C19H29ClN5O9P | 10MG
SDP
Supplier:  Medchemexpress LLC HY13697810MG
Encompass_Preferred
This compound is a potent, orally active CD73 (ecto-5'-nucleotidase, NT5E) inhibitor developed for cancer and immunology research. It blocks adenosine production to reverse immunosuppression and demonstrates low-nanomolar biochemical potency.
  • Potent CD73 inhibition with low-nanomolar biochemical potency.
  • Orally bioavailable in preclinical studies.
  • Reverses adenosine-mediated immunosuppression to enhance immune responses.
  • High reported purity suitable for research applications.
  • Available in small-quantity packages for screening and lead optimization.
Catalog No. 50-002-59499