Learn More
Cayman Chemical OP145trIfluoroACTAte salt 10mg
SDP
Supplier:  Cayman Chemical 3884110 MG
Encompass_Preferred
An antimicrobial peptide and a derivative of LL-37; active against E. coli, P. aeruginosa, C. albicans, and A. niger (MICs = 2, 3, 6, and 18 µM, respectively); PLGA microspheres containing OP-145 prevent biofilm formation in a mouse model of bone infection induced by surgical femoral fracture followed by bacterial inoculation at the fracture site
Catalog No. 50-291-8542