Learn More
Quanterix Inc Opal Rolaris 480 Reagent Pack
SDP
Supplier:  Quanterix Inc FP1500001KT
Encompass
Opal™ is a practical workflow for simultaneous detection of up to 6 tissue biomarkers plus nuclear counterstain, within a single image. The method is similar to standard immunohistochemistry and is accessible to many laboratories where standard IHC method development is performed. Standalone Opal Fluorophore reagent packs allow you to pick and choose your panel's biomarker quantity and design.
Catalog No. NC1685839