Learn More
MSC Viking High Visibility Vest, Orange, ANSI Class 2, 4X-Large and 5X-Large, 1 ea
Supplier:  MSC 42388694
Encompass
This high visibility vest is designed for public safety applications, providing enhanced visibility in various work environments. The vest is manufactured from a solid material in an orange color and features a zipper closure for secure wear.
  • Meets ANSI 107-2015 type R, class 2 standards
  • Complies with CSA class 2, level 2 requirements
  • Features 2 in. silver reflective tape
  • Includes 2 pockets, with 1 inside and 1 outside
  • Constructed from solid material
  • Machine washable for easy maintenance
  • Fits chest sizes 58 in. to 60 in.
Catalog No. 50-006-84557