Learn More
Sklar Instruments Operating Scissors, Sklar, 5 in., Sharp/Blunt, Straight, Smooth, Stainless Steel. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. EA/1.
SDP
Supplier:  Sklar Instruments 141050
Encompass_Preferred
Operating Scissors, Sklar, 5 in., Sharp/Blunt, Straight, Smooth, Stainless Steel. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is straight with smooth, sharp/blunt tips and a length of 5 inches.
Catalog No. 50-143-7364