Learn More
Cayman Chemical OphIobolIn A 5mg
SDP
Supplier:  Cayman Chemical 153815 MG
Encompass_Preferred
A sesterterpenoid fungal phytotoxin that binds and irreversibly antagonizes calmodulin, blocking the activation of calmodulin-dependent phosphodiesterase with an IC50 value of 9 μM
Catalog No. 50-297-0318