Learn More
Medchemexpress LLC HY-N3504 5mg Medchemexpress, Ophiopogonin D' CAS:65604-80-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN35045MG
Encompass_Preferred
Medchemexpress, HY-N3504 5mg Ophiopogonin D' CAS:65604-80-0 Ophiopogonin D', isolated from the tubers of Ophiopogon japonicus, is a rare naturally occurring C29 steroidal glycoside. Ophiopogonin D' shows cytotoxic activity against two human tumor cell lines MG-63 and SNU387 with IC50s of 3.09 μM and 3.63 μM, respectively. Ophiopogonin D' activates SIRT1 in a dose-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0413