Learn More
Medchemexpress LLC Ophiopogonone C | 477336-77-9 | 354.31 | 1 MG

Supplier: Medchemexpress LLC HYN119851MG
Ophiopogonone C is an isoflavone compound isolated from the ethanol extract of *Ophiopogon japonicus* tubers. It is for research use only.
- Molecular weight: 354.31
- Formula: C19H14O7
- Appearance: Solid, white to off-white
- Classification: Flavonoids, other flavonoids
- Initial source: Plants, Liliaceae, *Ophiopogon japonicus* (L. f.) Ker-Gawl.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.