Learn More
PATTERSON VETERINARY SUPPLY INC Patterson Veterinary - SurgiVet Adhesive Bandages, 1" x 3", Box of 100
Supplier:  PATTERSON VETERINARY SUPPLY INC 78946156
Encompass_Preferred
The VetOne VetPro Heartworm Antigen Test Kit is a rapid in-clinic test for the detection of Dirofilaria immitis antigen in dogs and cats. It uses a monoclonal antibody-based membrane to detect heartworm antigen in serum, plasma, or whole blood. The test provides results in 5-8 minutes and has a sensitivity of 98% and specificity of 100% in symptomatic dogs. This easy-to-use test requires only 3 drops of sample and includes everything needed to run the test. It allows veterinarians to quickly diagnose heartworm infection with an accurate in-house test.
Catalog No. NC2199806