Learn More
Selleck Chemical LLC OPN expression inhibitor 1 E4781-25MG
SDP
Supplier:  Selleck Chemical LLC E478125MG
Encompass_Preferred
OPN Expression Inhibitor 1 (Compound 11) is a DHA ether derivative containing a 1 2 3-triazole ring that effectively inhibits osteopontin (OPN) expression It exhibits significant potential as an anticancer agent by specifically targeting OPN a key factor in breast cancer progression and metastasis
Catalog No. 50-000-41998