Learn More
Selleck Chemical LLC Oprozomib
SDP
Supplier:  Selleck Chemical LLC S704950MG
Encompass_Preferred
Oprozomib is an orally bioavailable inhibitor for CT-L activity of 20S proteasome 5/LMP7 with IC50 of 36 nM/82 nM Phase 1/2
Catalog No. 50-000-02247