Learn More
TARGETMOL CHEMICALS INC OPROZOMIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T60415MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oprozomib (PR-047) (ONX 0912) an inhibitor for CT-L activity of 20S proteasome (beta)5(IC50=36 nM)/LMP7(IC50=82 nM) is orally bioavailable. Oprozomib also has potential antineoplastic activity. purity: 99%
Catalog No. 50-257-6618