Learn More
Medchemexpress LLC Oprozomib | 935888-69-0 | 99.8% | 100 MG

Supplier: Medchemexpress LLC HY12113100MG
Oprozomib is an orally bioavailable peptide epoxyketone proteasome inhibitor supplied for research use as a white to off-white solid with high purity and defined chemical identifiers.
- Orally bioavailable peptide epoxyketone proteasome inhibitor.
- Cas 935888-69-0 and molecular formula C25H32N4O7S.
- Molecular weight 532.61 g/mol.
- High purity (approximately 99.8%).
- Available as 100 MG solid for research use.
- Store powder at recommended low temperatures for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.