Learn More
Medchemexpress LLC Oprozomib | 935888-69-0 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1211350MG
Encompass_Preferred
Oprozomib (ONX 0912; PR-047) is an orally bioavailable, selective peptide epoxyketone proteasome inhibitor used in research to study proteasome inhibition, apoptosis induction, and related signaling pathways.
  • Orally bioavailable peptide epoxyketone proteasome inhibitor.
  • Selective inhibition of β5 and immunoproteasome LMP7 with reported IC50s ≈ 36 nM and 82 nM.
  • Induces apoptosis in multiple myeloma cells in preclinical studies.
  • High reported purity suitable for research applications.
  • Available in small research package sizes for laboratory use.
Catalog No. 50-002-32228