Learn More
Thomson Instrument Company Multiported Optimum Growth 5L Flask, Sterile
SDP
Supplier:  Thomson Instrument Company 931116PORTTRTF
Encompass_Preferred
Thomson’s Optimum Growth Multiport Flasks offer integrated ports that enable closed-system feeding, sampling, and media transfer—supporting sterile workflows without the need to open the flask, even during extended culture periods.
•Available in 125mL, 250mL, 500mL, 1.6L, 2.8L, 5L and 7L sizes.
•One-way sampling valve allows safe and aseptic sampling while the flask remains in the shaker or on the benchtop
•Two-way Luer lock port supports sterile feeding, inoculation, and direct media transfer to larger vessels
•Closed system design helps maintain sterility throughout use, reducing contamination risk and supporting GMP compliance
•Eliminates the need for cell bags up to 50L, reducing cost and complexity
•Compatible with multi-day batch feeding in shaker environments
•Includes sterile, pre-attached tubing for plug-and-play integration into cleanroom workflows
• Made in the USA
Catalog No. NC1412071