Learn More
Chemglass Life Sciences 8-Position Block for 28 x 83mm 10-20mL Round Bottom Microwave Vials, Two Tier Design
SDP
Supplier:  Chemglass Life Sciences OP660022
Encompass_Preferred
8-Position Block for 28 x 83mm 10-20mL Round Bottom Microwave Vials, Two Tier Design. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-874-142