Learn More
Chemglass Life Sciences 15-Position Block for 21 x 70mm 4 Dram (16mL) Flat Bottom Vials
SDP
Supplier:  Chemglass Life Sciences OP660009
Encompass_Preferred
15-Position Block for 21 x 70mm 4 Dram (16mL) Flat Bottom Vials. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-874-139