Learn More
Chemglass Life Sciences 11-Position Block for 28 x 70mm 40mL Scintillation & 28 x 95mm 60mL Scintillation Flat Bottom Vials, Two Tier Design
SDP
Supplier:  Chemglass Life Sciences OP660012
Encompass_Preferred
11-Position Block for 28 x 70mm 40mL Scintillation & 28 x 95mm 60mL Scintillation Flat Bottom Vials, Two Tier Design. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-877-460