Learn More
Optimize Technologies Optimize Universal Union for 1/16" OD Tubing, w/ Nuts & Ferrules, 10-32 Thread, .020" Thru , Stainless Steel
SDP
Supplier:  Optimize Technologies 102400373
Encompass
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi. The OPTI-LOK™ high pressure unions offer 10-32 threads for all high pressure applications with 1/16" OD tubing and provide zero dead volume connections with every use.
Catalog No. 50-113-420