Learn More
Bellco Glass, Inc. Optional Single Flow Station 5 Rotometers
SDP
Supplier:  Bellco Glass, Inc. 780510000
Encompass_Preferred
Optional Single Flow Station Rotometers. Rotometers for use with CellTrol II where gas addition is required. Typically used for mammalian cell culture where CO2 and N2 are used for pH control. Up to 5 rotometers are available. One required per vessel. Customer must supply flow rates from standard chart. BLENDS UP TO FIVE GASSES W/INDEPENDENT FLOW CONTROL.-8 3/4 X 15 X 8 3/4 (21.5 X 38 X 21.05 CM). -SST QUICK DISCONECT. UP TO FIVE SOLENOID VALVES (CUSTOMER SPECIFIED). INDEPENDENT HEADSPACE OVERLAY AND GAS SPARGE. MADE IN THE USA
Catalog No. 50-194-6356