Learn More
Electron Microscopy Sciences Digital Microscopes Model AF4915ZTL, 1/EA
SDP
Supplier:  Electron Microscopy Sciences WF20
Encompass_Preferred
Adding the optional WF-20 wireless streamer connection to the Dino-Lite Edge AF4915ZTL, which features a long battery life and stable WiFi image transmission, makes the Dino-Lite a completely cordless and portable microscope.
Catalog No. 50-209-1622