Learn More
Electron Microscopy Sciences Optivison™ Optivisor Auxilary Lens
SDP
Supplier:  Electron Microscopy Sciences 6810750
Encompass_Preferred
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Increase 2-1/2X magnification
Catalog No. 50-305-20