Learn More
Beckman Coulter OptiXTRACT Adapter Kit - 50 adapters for syringe-to-needle connections and tower mounting
Supplier:  Beckman Coulter D18845
Encompass_Preferred
The Beckman Coulter OptiXTRACT Adapter Kit contains 50 adapters used to connect a syringe to the needle and to position the syringe/needle assembly on the OptiXTRACT tower for density gradient sample recovery.
  • Contains 50 single-use adapters.
  • Enables secure syringe-to-needle connection for controlled sample extraction.
  • Supports consistent placement of the syringe/needle assembly on the instrument tower.
  • Reduces risk of sample contamination through single-use components.
  • Compatible with standard syringe and needle configurations used for gradient recovery.
Catalog No. NC3920958