Learn More
Aviva Systems Biology Corp OR2M5 Antibody - C-terminal region (ARP59741_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59741_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2M5. It was validated on Western Blot. . Alternate names: OR2M5PApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-286-803