Learn More
Med Vet International Oral Blunt Sterile Cannula - 13G x 6cm, 2/Pack, J1621A
Supplier:  Med Vet International JOR1621A
Encompass_Preferred
Atraumatic Blunt Tip Cannulas. JorVet is offering a range of blunt tip needles and Olive tip cannulas for a variety of clinical applications where a sharp point is not suitable or desired. These blunt needles come in a variety of sizes from small to large plus different materials from malleable polypropylene to more ridid polymer. All have female luer lock fittings to readily pair with standard syringes. • Feeding needles for oral administration of liquids • Wound flushing/irrigation • Anal sac infusion • Male cat urethral flush • Lacrimal duct irrigation Atraumatic Blunt Tip Cannulas - Oral Blunt Sterile Cannula - 13G x 6cm 2 Pk
Catalog No. 50-005-83171