Learn More
Medchemexpress LLC Orally active dual dUTPase/dihydropyrimidine dehydrogenase (DPD) inhibitor | 1198221-21-4 | 99.9% | 451.54 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12406250MG
Encompass_Preferred
TAS-114 is an orally active dual dUTPase/dihydropyrimidine dehydrogenase (DPD) inhibitor, which can improve the therapeutic efficacy of fluoropyrimidine.
  • Orally active
  • Dual dUTPase/dihydropyrimidine dehydrogenase (DPD) inhibitor
  • Can improve the therapeutic efficacy of fluoropyrimidine
Catalog No. 50-004-37965