Learn More
McMaster-Carr ORANGE LID VAC GLASS BOTTLE
Supplier:  McMaster-Carr 4787T73
Encompass
McMaster-Carr offers a wide selection of glass bottles in various sizes, shapes, and colors. The bottles are made from durable borosilicate glass and feature threaded openings for secure closure. Popular options include Boston round bottles, reagent bottles, media bottles, and wide mouth jars. The bottles have volume capacities ranging from 30mL to 4L. They can be used for storing liquids, powders, and small items. The threaded openings accept caps, droppers, and various closure types for an airtight seal. With the variety of sizes and styles, these glass bottles have uses across many industries and applications.
Catalog No. NC2874989