Learn More
Elemental Scientific Inc ORANGED/GREEN FLARED TUB 12/P
SDP
Supplier:  Elemental Scientific Inc ES43971038
Encompass_Preferred
Peristaltic Pump Tubing, PVC, 2-stop, 0.38mm ID, 2.0mm flared 151mm bridge distance (orange/green), 12 pc.
Catalog No. NC9049152