Learn More
Medchemexpress LLC Orbifloxacin | 113617-63-3 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB091510MM/1ML
Encompass_Preferred
Orbifloxacin is an orally administrable antibiotic that disrupts the replication and proliferation of bacterial DNA, inhibits bacterial growth, and exerts bactericidal activity. It inhibits the growth of canine-derived E. coli and Pseudomonas aeruginosa isolates and is used in research related to bacterial infections.
  • Orally administrable antibiotic.
  • Disrupts the replication and proliferation of bacterial DNA.
  • Inhibits bacterial growth and exerts bactericidal activity.
  • Inhibits the growth of canine-derived E. coli and Pseudomonas aeruginosa isolates.
  • Used in research related to bacterial infections.
Catalog No. 50-004-08693