Learn More
Medchemexpress LLC Orbifloxacin | 113617-63-3 | 99.8% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0915200MG
Encompass_Preferred
Orbifloxacin is an orally administrable antibiotic that disrupts the replication and proliferation of bacterial DNA, inhibiting bacterial growth and exerting bactericidal activity. It has been shown to inhibit the growth of canine-derived *E. coli* and *Pseudomonas aeruginosa* isolates, and is used in research related to bacterial infections.
  • Antibiotic mechanism: Disrupts bacterial DNA replication and proliferation, leading to bactericidal activity.
  • Target: Quinolone.
  • Inhibits the growth of *Piscirickettsia salmonis* LF-89 VR-1361 (0.015-4.0 μg/mL) with sustained effect over 7 days.
  • Induces post-antibiotic effect (PAE) and post-antibiotic sub-MIC effect (PA-SME) in clinical canine isolates.
  • Exhibits concentration-dependent bactericidal activity against *Staphylococcus aureus* and time-dependent bactericidal activity against *Staphylococcus intermedius*.
Catalog No. 50-004-09234