Learn More
Cayman Chemical OrbIfloxacIn 250mg
SDP
Supplier:  Cayman Chemical 17353250 MG
Encompass_Preferred
A fluoroquinolone antibiotic used in animals, including dogs, cattle, and swine, to combat gastrointestinal and respiratory infections
Catalog No. 50-296-6078