Learn More
USA Scientific Inc Orbital-Genie Ratchet Clamps
Supplier:  USA Scientific Inc 74001500
Encompass
Orbital-Genie orbital shaker with ratchet clamps; measures 20 in wide x 23 in deep. 120 V.
Catalog No. 50-274-4802