Learn More
TARGETMOL CHEMICALS INC ORELABRUTINIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T123175MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Orelabrutinib (ICP-022) is an orally active and irreversible inhibitor of Bruton's tyrosine kinase (BTK). purity: 99%
Catalog No. 50-258-1468