Learn More
Medchemexpress LLC HY-129390 5mg Medchemexpress, Orelabrutinib CAS:1655504-04-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1293905MG
Encompass_Preferred
Medchemexpress, HY-129390 5mg Orelabrutinib CAS:1655504-04-3 Orelabrutinib (ICP-022) is a potent, orally active, and irreversible Bruton's tyrosine kinase (BTK) inhibitor with potential antineoplastic activity. Orelabrutinib prevents both the activation of the B-cell antigen receptor (BCR) signaling pathway and BTK-mediated activation of downstream survival pathways, inhibiting the growth of malignant B-cells that overexpress BTK[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0155