Learn More
TARGETMOL CHEMICALS INC Orellanine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T338171MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Orellanine is a nephrotoxic compound extracted from the mushroom Cortinarius orellanus. Orellanine strongly inhibits the synthesis of macromolecules (Proteins, RNA and DNA) in Madin-Darby canine kidney cells and in rat liver mitochondria. Purity 90%
Catalog No. 50-287-6330