Learn More
Cayman Chemical OrexIn A amIdebovInhumn 1mg
SDP
Supplier:  Cayman Chemical 150731 MG
Encompass_Preferred
A 33 amino acid hypothalamic neuropeptide with a role in appetite regulation, wakefulness, locomotor activity, sympathetic and HPA activity, and pain thresholds; binds the orexin-1 and -2 receptors with equal affinity
Catalog No. 50-297-1569