Learn More
Medchemexpress LLC Orexin A human rat 1mg | 205640-90-0 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1062241MG
Encompass_Preferred
Orexin A (human rat mouse) (Hypocretin-1 (human rat mouse)) a 33 amino acid excitatory neuropeptide orchestrates diverse central and peripheral processes Orexin A (human rat mouse) binds and activates two types of G protein-coupled receptors the orexin-1 receptor (OX1R) and the orexin-2 receptor (OX2R) Orexin A (human rat mouse) has a role in the regulation of feeding behavior Orexin A (human rat mouse) is an effective anti-nociceptive and anti-hyperalgesic agent in mice and rats[1 [2
Catalog No. 50-002-41819