Learn More
Sigma Aldrich Fine Chemicals Biosciences Orexin A human rat mouse 91MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences O6012.1MG
Encompass
Orexin A also known as hypocretin 1 (hcrt1) is a 33 amino acid excitatory neuropeptide localized in a small set of neurons in the perifornical area of the hypothalamus. It is synthesized from the precursor protein prepro-orexin in neurons. Orexin A binds to its G-protein-coupled receptor orexin-1 (OX1) which is also called as hcrt receptors 1 (hcrtr1).
Catalog No. 50-290-2286