Learn More
Cayman Chemical OrexIn A17-33humnmous 500ug
SDP
Supplier:  Cayman Chemical 36765500 UG
Encompass_Preferred
A peptide orexin receptor 1 agonist; selectively induces calcium mobilization in CHO cells overexpressing human OX1R over CHO cells overexpressing human OX2R (EC50s = 8.29 and 187 nM, respectively)
Catalog No. 50-291-3320