Learn More
Medchemexpress LLC HY-118152 10mg Medchemexpress, Org-12962 CAS:132834-56-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11815210MG
Encompass_Preferred
Medchemexpress, HY-118152 10mg Org-12962 CAS:132834-56-1 Org-12962 is a potent, selective and orally active 5-HT2C receptor agonist with a pEC50 value of 7.01. Org-12962 also exhibits high effacy for the 5-HT2A and 5-HT2B receptor with pEC50s of 6.38 and 6.28, respectively[3].Org-12962 displays antiaversive effects in a rat model of panic-like anxiety. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9498