Learn More
Medchemexpress LLC Org 274179-0 | 1421683-12-6 | C28H27F3N2O2 | 25 MG

Supplier: Medchemexpress LLC 25MGHY153213
Org 274179-0 is a potent and allosteric antagonist of the thyroid-stimulating hormone (TSH) receptor, with an IC50 of nanomolar concentration. It completely inhibits TSH (and TSI)-mediated TSH receptor activation with minimal impact on the potency of TSH. This product is suitable for research related to Graves' disease (GD).
- Potent and allosteric antagonist of TSH receptor
- Completely inhibits TSH (and TSI)-mediated TSH receptor activation
- Minimal impact on TSH potency
- Suitable for Graves' disease research
- Purity: 98.63%
- Appearance: solid
- Color: light yellow to yellow
- Solubility in DMSO: 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.