Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Org41841 | 301847-37-0 | 99.8% | 402.53 g/mol | C19H22N4O2S2 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY10027125MG

Encompass_Preferred

Org41841 is a small-molecule partial agonist of the luteinizing hormone/chorionic gonadotropin receptor (LHCGR) and the thyroid-stimulating hormone receptor (TSHR), used for receptor pharmacology research. It exhibits EC50 values of 0.2 μM (LHCGR) and 7.7 μM (TSHR).

  • Partial agonist at LHCGR and TSHR with EC50s of 0.2 μM and 7.7 μM.
  • High purity (99.77%) appropriate for biochemical assays.
  • Molecular weight 402.53 g/mol.
  • Chemical formula C19H22N4O2S2.
  • CAS number 301847-37-0 for unambiguous identification.
  • Available in multiple milligram quantities and DMSO solution formats for screening.
  • Intended for research use only and not for human or clinical use.

Catalog No. 50-002-14241


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.