Learn More
ORGANABIO LLC Human Peripheral Blood Mononuclear Cells (PBMCs) / Cryopreserved / Single Donor / 30-45x vials of 1x10E8 cells/vial / OrganaBio LEUKOPAC - PBMC / Research Use Only
Supplier:  ORGANABIO LLC PBMCPB100HB
Encompass
Human peripheral blood mononuclear cells (PBMCs) from recallable / HLA-typed healthy adult donors isolated by density gradient centrifugation. Comprehensive characterization including immunophenotyping (CD45 / CD14 / CD19 / CD56 / CD3 / CD3CD4 / CD3CD8) / infectious disease testing / and high-resolution HLA typing (Class I and II). Half leukopak processed into vials. Availble in 1X10E8 cells/vial. OrganaBio Human PBMCs are isolated from peripheral blood of normal / healthy adult donors by density gradient centrifugation. Each lot is cryopreserved and characterized by immunophenotyping. Comprehensive donor characterization includes infectious disease testing panel and high-resolution HLA typing with full donor demographics (age / sex / BMI / race / ethnicity / blood type).
Catalog No. NC3997822